DL-Lithium lactate 99%, Extra Pure

Supplier: Thermo Scientific Chemicals

Synonyms: DL-Lactic acid lithium salt, (±)-2-Hydroxypropanoic acid lithium salt, Lithium 2-hydroxypropanoate, (±)-2-Hydroxypropionic acid lithium salt, (±)-Lactic acid lithium salt

AC41333-0050 AC41333-0250 AC41333-1000 AC41333-5000
TS41333-0050EA 38 USD
TS41333-0050 TS41333-0250 TS41333-1000 TS41333-5000
DL-Lithium lactate 99%, Extra Pure
DL-Lithium lactate
Formula: H₃CCH(OH)COOLi
MW: 96.01 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00065399
CAS Number: 867-55-0
EINECS: 212-761-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Solubility (10% in water) Clear colorless
pH 6 to 9 (1% solution)
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR