tri-Sodium citrate dihydrate ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

AC44633-0010 AC44633-0050 AC44633-2500
TS44633-0010EA 74 USD
TS44633-0010 TS44633-0050 TS44633-2500
tri-Sodium citrate dihydrate ACS
tri-Sodium citrate dihydrate
Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
pH 7.0 to 9.0 (5 % in water at 25.0°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤0.003 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %
Ammonia (NH3) ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR