1-Dodecansulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Supplier: Thermo Scientific Chemicals
AC39672-0050
AC39672-0250
TS39672-0050EA
120
USD
TS39672-0050
TS39672-0250
1-Dodecansulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
1-Dodecansulphonic acid sodium salt
Formula:
C₁₂H₂₅NaO₃S Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007527 CAS Number: 2386-53-0 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % |
UV | at 230 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.18 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...