3'-Bromoacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: m-Bromoacetophenone

AC10658-0050 AC10658-0250 AC10658-1000
TS10658-0050EA 14 USD
TS10658-0050 TS10658-0250 TS10658-1000
3'-Bromoacetophenone 97%
3'-Bromoacetophenone
Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 79…81 °C (2 mmHg)
Density: 1.49 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000083
CAS Number: 2142-63-4
EINECS: 218-396-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5749 to 1.5769 (20°C, 589 nm)
GC ≥96.0 %
Residue after ignition ≤0.3 %
Specific optical rotation -45° to -51° (20°C, 589 nm) (c=10, 75% acetic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR