Diethyl sulfate 99%

Supplier: Thermo Scientific Chemicals
Danger

AC11456-0010 AC11456-0025 AC11456-0100 AC11456-0250
TS11456-0010EA 58 USD
TS11456-0010 TS11456-0025 TS11456-0100 TS11456-0250
Diethyl sulfate 99%
Diethyl sulphate
Formula: C₄H₁₀O₄S
MW: 154.19 g/mol
Boiling Pt: 208 °C (760 mmHg)
Melting Pt: –24 °C
Density: 1.1795 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00009099
CAS Number: 64-67-5
EINECS: 200-589-6
UN: 1594
ADR: 6.1,II
Merck Index: 13,03156

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4010 (20°C, 589 nm)
Water ≤0.15 %
GC ≥98.5 %
Color scale ≤15 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR