Calcium sulphate hemihydrate 97+%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Plaster of Paris

AC38535-0025 AC38535-1000 AC38535-5000
TS38535-0025EA 114 USD
TS38535-0025 TS38535-1000 TS38535-5000
Calcium sulphate hemihydrate 97+%, pure
Calcium sulphate hemihydrate
Formula: CaSO₄·0.5H₂O
MW: 145.15
Density: 2.73
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 10034-76-1
EINECS: 231-900-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Titration Complexometric ≥97.0 % (On dry substance)
Identification Confirmed
Loss on ignition 4.5 % to 8.0 % (500°C, 2 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR