Potassium sulfate, anhydrous 99+%, powder for analysis

Supplier: Thermo Scientific Chemicals

AC20777-0050 AC20777-5000
TS20777-0050EA 164 USD
TS20777-0050 TS20777-5000
Potassium sulfate, anhydrous 99+%, powder for analysis
Potassium sulphate
Formula: K₂SO₄
Boiling Pt: 1689 °C (760 mmHg)
Melting Pt: 1067 °C
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Appearance White Powder or crystals
pH 5.5 to 8 (5 % at 20°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤5 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤20 ppm
Sodium (Na) ≤200 ppm
Heavy metals (as Pb) ≤5 ppm
Total nitrogen (as N) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR