Antimony(V) sulphide, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Antimony pentasulphide, Diantimony pentasulphide

AC33041-0010 AC33041-0500 AC33041-2500
TS33041-0010EA 218 USD
TS33041-0010 TS33041-0500 TS33041-2500
Antimony(V) sulphide, pure
Antimony(V) sulphide
Formula: S₅Sb₂
MW: 403.85
Density: 4.12
Storage Temperature: Ambient
MDL Number: MFCD00016321
CAS Number: 1315-04-4
EINECS: 215-255-5
UN: 3179
ADR: 4.1 II
Merck Index: 12,00738

Order Now

Specification Test Results

Appearance Yellow to orange-red Powder
Assay ≥54 % (Sb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR