Calcium hydrogen orthophosphate dihydrate 98+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Calcium phosphate dibasic dihydrate, Calcium hydrogen phosphate dihydrate, Calcium hydrogenphosphate dihydrate, Dibasic Calcium Phosphate Dihydrate

AC38983-0010 AC38983-0050
TS38983-0010EA 85 USD
TS38983-0010 TS38983-0050
Calcium hydrogen orthophosphate dihydrate 98+%, Extra Pure
Calcium hydrogen orthophosphate dihydrate
Formula: HCaO₄P·2H₂O
MW: 172.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149621
CAS Number: 7789-77-7

Order Now

Specification Test Results

Appearance White to almost white Powder
Titration Complexometric 98.0 to 105.0 % (on dried substance)
Identification Phosphate: positive
Loss on ignition 24.5 % to 26.5 % (800°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR