Calcium hydroxide 99.995% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC38561-0050 AC38561-0250
TS38561-0050EA 78.8 USD
TS38561-0050 TS38561-0250
Calcium hydroxide 99.995% (trace metals basis)
Calcium hydroxide
Formula: Ca(OH)₂
Boiling Pt: 2850 °C
Melting Pt: 580 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0

Order Now

Specification Test Results

Appearance White Powder or crystals
Titration Complexometric ≥54 % (Ca)
Total trace metal impurities ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR