Barium hydroxide octahydrate 98+% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC20314-0010 AC20314-0050 AC20314-2500
TS20314-0010EA 93 USD
TS20314-0010 TS20314-0050 TS20314-2500
Barium hydroxide octahydrate 98+% for analysis
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Boiling Pt: 780 °C (1013 hPa)
Melting Pt: 78 °C
Density: 2.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,II
Merck Index: 13,00980

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or flakes
Titration with HCl ≥98.0 %
Chloride (Cl) ≤20 ppm
Barium carbonate ≤2 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Strontium (Sr) ≤1.5 %
Sulfur (S) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 % (in HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR