Titanium(IV) butanolate 98+%

Supplier: Thermo Scientific Chemicals
Danger

AC22319-0010 AC22319-0020 AC22319-0100 AC22319-2500 AC22319-5000
TS22319-0010EA 142 USD
TS22319-0010 TS22319-0020 TS22319-0100 TS22319-2500 TS22319-5000
Titanium(IV) butanolate 98+%
Titanium(IV) butanolate
Formula: C₁₆H₃₆O₄Ti
Boiling Pt: 310…314 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.99 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00009433
CAS Number: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Viscosity 50 to 90 cP (at 25°C)
Gravimetry 23.0 to 23.9 % (TiO2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR