Tetraethyl orthosilicate (TEOS) 98%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetraethoxysilane

AC42036-0010 AC42036-1000
TS42036-0010EA 84 USD
TS42036-0010 TS42036-1000
Tetraethyl orthosilicate (TEOS) 98%, AcroSeal™
Tetraethyl orthosilicate (TEOS)
Formula: Si(OC₂H₅)₄
MW: 208.33 g/mol
Boiling Pt: 168
Melting Pt: -85.5
Density: 0.94 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00009062
CAS Number: 78-10-4
UN: 1292
ADR: 3 III
Merck Index: 13,03882

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3820 to 1.3840 (20°C, 589 nm)
GC ≤1 % (ethanol)
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR