Trimethoxysilane 95%

Supplier: Thermo Scientific Chemicals
Danger

AC39217-0050 AC39217-0250 AC39217-1000
TS39217-0050EA 59 USD
TS39217-0050 TS39217-0250 TS39217-1000
Trimethoxysilane 95%
Trimethoxysilane
Formula: C₃H₁₀O₃Si
MW: 122.2 g/mol
Boiling Pt: 81…83 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.96 g/cm³
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00008340
CAS Number: 2487-90-3
EINECS: 219-637-2
UN: 3384
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3570 to 1.3600 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR