Lead(II,IV) oxide 98%

Supplier: Thermo Scientific Chemicals
Danger

AC22111-0050 AC22111-0051 AC22111-1000 AC22111-5000
TS22111-0050EA 21 USD
TS22111-0050 TS22111-0051 TS22111-1000 TS22111-5000
Lead(II,IV) oxide 98%
Lead(II,IV) oxide
Formula: Pb₃O₄
Boiling Pt: 2682 °C (1013 hPa)
Melting Pt: 500 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00078491
CAS Number: 1314-41-6
EINECS: 215-235-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,05449

Order Now

Specification Test Results

Appearance Orange Powder
Assay ≥97.5 % (PB3O4)
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤100 ppm
Manganese (Mn) ≤10 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤100 ppm
Zinc (Zn) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR