Potassium hexacyanoferrate(III) 99+% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

AC22311-0025 AC22311-1000 AC22311-5000
TS22311-0025EA 343 USD
TS22311-0025 TS22311-1000 TS22311-5000
Potassium hexacyanoferrate(III) 99+% for analysis
Potassium hexacyanoferrate(III)
Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Specification Test Results

Appearance Orange to red Crystals and/or chunks
Titration Iodometric ≥99.0 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Hexacyanoferrate II ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR