Disodium oxalate 98.5%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances

AC20772-0025 AC20772-1000 AC20772-5000
TS20772-0025EA 298 USD
TS20772-0025 TS20772-1000 TS20772-5000
Disodium oxalate 98.5%, extra pure
Disodium oxalate
Formula: Na₂C₂O₄
Melting Pt: 250…270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723

Order Now

Specification Test Results

Appearance White Powder
Loss on drying ≤0.1 % (105°C, 2 h)
Titration with KMnO4 ≥98.0 %
Insoluble matter ≤0.2 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR