Zinc acetate dihydrate 97%, meetSpec_USP

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

AC45188-0010 AC45188-0025 AC45188-0100
TS45188-0010EA 75 USD
TS45188-0010 TS45188-0025 TS45188-0100
Zinc acetate dihydrate 97%, meetSpec_USP
Zinc acetate dihydrate
Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 237
Density: 1.84 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
UN: 3077
ADR: 9 III
Merck Index: 12,10256

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric 98.0 to 102.0 %
pH 6.0 to 8.0 (5 % solution)
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.01 %
Arsenic (As) ≤3 ppm
Lead (Pb) ≤0.002 %
Insoluble matter ≤0.005 %
Alkalies + earths ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR