Zinc acetate dihydrate 98+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC31721-0025
AC31721-5000
TS31721-0025EA
170
USD
TS31721-0025
TS31721-5000
Zinc acetate dihydrate 98+% ACS
Zinc acetate dihydrate
Formula:
(H₃CCOO)₂Zn·2H₂O MW: 219.51 g/mol Melting Pt: 237 °C Density: 1.84 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00066961 CAS Number: 5970-45-6 EINECS: 209-170-2 UN: 3077 ADR: 9,III Merck Index: 12,10256 |
Specification Test Results
Appearance | White Crystals |
Titration Complexometric | 98.0 to 101.0 % |
pH | 6.0 to 7.0 (5 % at 25°C) |
Chloride (Cl) | ≤5 ppm |
Sulfate (SO4) | ≤0.005 % |
Antimony (Sb) | ≤2 ppm |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤5 ppm |
Lead (Pb) | ≤0.002 % |
Magnesium (Mg) | ≤0.005 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.05 % |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...