Copper(II) acetate, anhydrous 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

AC40537-0010 AC40537-0500 AC40537-2500
TS40537-0010EA 1041 USD
TS40537-0010 TS40537-0500 TS40537-2500
Copper(II) acetate, anhydrous 99%, pure
Copper(II) acetate
Formula: CuCH₃CO₂
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008690
CAS Number: 142-71-2
EINECS: 205-553-3
UN: 3260
ADR: 8,II
Merck Index: 12,02690

Order Now

Specification Test Results

Appearance Green to blue Crystalline powder
Infrared spectrum Conforms
Titration with Na2S2O3 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR