Cesium acetate 99% for analysis

Supplier: Thermo Scientific Chemicals
Warning

AC39083-0250 AC39083-1000 AC39083-5000
TS39083-0250EA 74 USD
TS39083-0250 TS39083-1000 TS39083-5000
Cesium acetate 99% for analysis
Cesium acetate
Formula: H₃CCOOCs
Melting Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00013056
CAS Number: 3396-11-0

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤5 ppm
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤10 ppm
Calcium (Ca) ≤5 ppm
Chromium (Cr) ≤0.5 ppm
Iron (Fe) ≤0.5 ppm
Lithium (Li) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤0.5 ppm
Potassium (K) ≤5 ppm
Sodium (Na) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR