Cesium acetate 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid cesium salt

AC19104-0250 AC19104-1000
TS19104-0250EA 68 USD
TS19104-0250 TS19104-1000
Cesium acetate 99%, pure
Cesium acetate
Formula: H₃CCOOCs
MW: 191.95 g/mol
Melting Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR