Dibutyl sebacate 93%

Supplier: Thermo Scientific Chemicals

Synonyms: Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate

AC40665-0010 AC40665-0025 AC40665-2500
TS40665-0010EA 111 USD
TS40665-0010 TS40665-0025 TS40665-2500
Dibutyl sebacate 93%
Dibutyl sebacate
Formula: C₁₈H₃₄O₄
MW: 314.47 g/mol
Boiling Pt: 180
Melting Pt: -11
Density: 0.94 g/cm³
Flash Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00027218
CAS Number: 109-43-3
EINECS: 203-672-5

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥91 %
Color scale ≤40 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR