Silver carbonate 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

AC17697-0250 AC17697-1000
TS17697-0250EA 132 USD
TS17697-0250 TS17697-1000
Silver carbonate 99%, pure
Silver carbonate
Formula: CAg₂O₃
MW: 275.75 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003403
CAS Number: 534-16-7
EINECS: 208-590-3
UN: 3077
Merck Index: 12,08650

Order Now

Specification Test Results

Appearance Yellow to green Granular powder
Assay ≥98.5 %
Melting point 176 to 180 °C
Water ≤1 %
HPLC ≥97.5 %
Specific optical rotation +10° to +12° (20 °C, 589 nm) (5 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR