Sodium hypochlorite 10 - 15% available chlorine in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC21925-0025
AC21925-0100
AC21925-5000
TS21925-0025EA
140
USD
TS21925-0025
TS21925-0100
TS21925-5000
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution
Sodium hypochlorite
Formula:
ClNaO MW: 74.44 g/mol Melting Pt: –16 °C Density: 1.2 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00011120 CAS Number: 7681-52-9 EINECS: 231-668-3 UN: 1791 ADR: 8,III |
Specification Test Results
Appearance | Clear yellow Solution |
Titration Iodometric | 10 to 15 % |
Iron (Fe) | ≤5 ppm |
Free alkali (NaOH) | ≤1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...