(3-Chloropropyl)trimethoxysilane 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Chloropropyltrimethoxysilane, 3-(Trimethoxysilyl)propylchloride, Trimethoxy(3-chloropropyl)silane

AC13728-0250 AC13728-1000 AC13728-5000
TS13728-0250EA 26 USD
TS13728-0250 TS13728-1000 TS13728-5000
(3-Chloropropyl)trimethoxysilane 98+%
(3-Chloropropyl)trimethoxysilane
Formula: ClH₂CCH₂CH₂Si(OCH₃)₃
MW: 198.72 g/mol
Boiling Pt: 195…196 °C (750 mmHg)
Density: 1.07 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00000997
CAS Number: 2530-87-2
EINECS: 219-787-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20°C, 589 nm)
GC ≥98.0 %
Specific optical rotation +16° to +20° (20°C, 589 nm) (c=5% in Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR