Cadmium(II) chloride hemipentahydrate 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19702-0250
AC19702-2500
TS19702-0250EA
37
USD
TS19702-0250
TS19702-2500
Cadmium(II) chloride hemipentahydrate 99+%
Cadmium(II) chloride hemipentahydrate
Formula:
CdCl₂·2.5H₂O MW: 228.35 g/mol Melting Pt: 34 °C |
MDL Number:
MFCD00799071 CAS Number: 7790-78-5 EINECS: 233-296-7 UN: 2570 ADR: 6.1,III Merck Index: 13,01617 |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or chunks |
Titration Complexometric | 79.5 to 81.0 % (CdCl2) |
Nitrate and Nitrite (as NO3) | ≤0.003 % |
Sulfate (SO4) | ≤50 ppm |
Ammonium (NH4) | ≤50 ppm |
Copper (Cu) | ≤5 ppm |
Iron (Fe) | ≤5 ppm |
Lead (Pb) | ≤50 ppm |
Zinc (Zn) | ≤500 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...