Fentin chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chlorotriphenylstannane, Triphenyltin chloride

AC31729-0050 AC31729-1000 AC31729-5000
TS31729-0050EA 22 USD
TS31729-0050 TS31729-1000 TS31729-5000
Fentin chloride 95%
Fentin chloride
Formula: C₁₈H₁₅ClSn
MW: 385.48 g/mol
Boiling Pt: 397
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00000519
CAS Number: 639-58-7
EINECS: 211-358-4
UN: 3146
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 108 °C
Titration Argentometric ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR