Triisopropylchlorosilane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chlorotriisopropylsilane, Triisopropylsilyl chloride, Triisopropylchlorosilane

AC22126-0100 AC22126-0500 AC22126-2500
TS22126-0100EA 79 USD
TS22126-0100 TS22126-0500 TS22126-2500
Triisopropylchlorosilane 97%
Triisopropylchlorosilane
Formula: C₉H₂₁ClSi
MW: 192.8 g/mol
Boiling Pt: 198 °C (760 mmHg)
Density: 0.902 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00009656
CAS Number: 13154-24-0
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR