Dimethyldichlorosilane 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dichloro(dimethyl)silane, Dichlorodimethylsilane

AC43088-1000 AC43088-8000
TS43088-1000EA 38 USD
TS43088-1000 TS43088-8000
Dimethyldichlorosilane 99+%, AcroSeal™
Dimethyldichlorosilane
Formula: C₂H₆Cl₂Si
MW: 129.06 g/mol
Boiling Pt: 70
Melting Pt: -76
Density: 1.06 g/cm³
Flash Pt: –9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000491
CAS Number: 75-78-5
UN: 1162
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4030 to 1.4060 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR