Iron(II) chloride, anhydrous 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron dichloride, Ferrous chloride

AC38935-0250 AC38935-2500
TS38935-0250EA 104 USD
TS38935-0250 TS38935-2500
Iron(II) chloride, anhydrous 97%
Iron(II) chloride
Formula: Cl₂Fe
MW: 126.75 g/mol
Boiling Pt: 2023
Melting Pt: 674
Storage Temperature: Ambient
MDL Number: MFCD00011004
CAS Number: 7758-94-3
UN: 3260
ADR: 8 III
Merck Index: 12,04091

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Titration with KMnO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR