Mercury(I) chloride ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimercury dichloride, Mercurous chloride, Calomel
AC44759-1000
AC44759-5000
TS44759-1000EA
107
USD
TS44759-1000
TS44759-5000
Mercury(I) chloride ACS
Mercury(I) chloride
Formula:
Cl₂Hg₂ MW: 472.09 g/mol Boiling Pt: 384 Melting Pt: 383 (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00011043 CAS Number: 10112-91-1 UN: 2025 ADR: 6.1 III Merck Index: 12,05951 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Titration Iodometric | ≥99.5 % |
Sulfate (SO4) | ≤0.01 % |
Mercuric Chloride | ≤0.01 % (HgCl2) |
Residue after reduction | ≤0.02 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...