Mercury(I) chloride 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dimercury dichloride, Mercurous chloride, Calomel

AC21312-0050 AC21312-1000 AC21312-5000
TS21312-0050EA 19 USD
TS21312-0050 TS21312-1000 TS21312-5000
Mercury(I) chloride 99+%, extra pure
Mercury(I) chloride
Formula: Cl₂Hg₂
MW: 472.09 g/mol
Boiling Pt: 384
Melting Pt: 383 (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011043
CAS Number: 10112-91-1
EINECS: 233-307-5
UN: 2025
ADR: 6.1 III
Merck Index: 12,05951

Order Now

Specification Test Results

Appearance Off-white Powder
Titration Iodimetric ≥99.0 %
Mercuric Chloride ≤0.01 % (HgCl2)
Residue after reduction ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR