Copper(I) chloride 90+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cuprous chloride, Copper chloride

AC40587-1000 AC40587-5000
TS40587-1000EA 82 USD
TS40587-1000 TS40587-5000
Copper(I) chloride 90+% ACS
Copper(I) chloride
Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1490 °C (760 mmHg)
Melting Pt: 430 °C
Density: 4.14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8,III
Merck Index: 13,02690

Order Now

Specification Test Results

Appearance White to light green Crystalline powder
Titration with KMnO4 ≥90.0 %
Sulfate (SO4) ≤1000 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤50 ppm
Potassium (K) ≤200 ppm
Sodium (Na) ≤500 ppm
Insoluble matter ≤0.02 % (in acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR