Copper(I) chloride 97+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cuprous chloride, Copper chloride

AC27052-0010 AC27052-0025 AC27052-5000
TS27052-0010EA 117 USD
TS27052-0010 TS27052-0025 TS27052-5000
Copper(I) chloride 97+%, extra pure
Copper(I) chloride
Formula: ClCu
Boiling Pt: 1490 °C (760 mmHg)
Melting Pt: 430 °C
Density: 4.14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8,III
Merck Index: 13,02690

Order Now

Specification Test Results

Appearance Light green to gray Crystalline powder
Titration Complexometric 97.0 to 103.0 %
Sulfate (SO4) ≤500 ppm
Arsenic (As) ≤5 ppm
Iron (Fe) ≤100 ppm
Insoluble matter ≤0.05 % (in acid)
Subst nt pptd by H2S(SO4) ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR