Copper(I) chloride 99.99% (trace metals basis), Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cuprous chloride, Copper chloride
AC21242-0100
AC21242-1000
TS21242-0100EA
90
USD
TS21242-0100
TS21242-1000
Copper(I) chloride 99.99% (trace metals basis), Extra Pure
Copper(I) chloride
Formula:
ClCu MW: 99 g/mol Boiling Pt: 1367 °C (1013 hPa) Melting Pt: 430 °C Density: 4.14 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010971 CAS Number: 7758-89-6 EINECS: 231-842-9 UN: 2802 ADR: 8,III Merck Index: 13,02690 |
Specification Test Results
Appearance | White to light green Crystalline powder |
Total trace metal impurities | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...