Copper(I) chloride 97%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cuprous chloride, Copper chloride
AC19964-0025
AC19964-0250
AC19964-5000
TS19964-0025EA
244
USD
TS19964-0025
TS19964-0250
TS19964-5000
Copper(I) chloride 97%, pure
Copper(I) chloride
Formula:
ClCu MW: 99 g/mol Boiling Pt: 1367 Melting Pt: 430 Density: 4.14 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010971 CAS Number: 7758-89-6 EINECS: 231-842-9 UN: 2802 ADR: 8 III Merck Index: 13,02690 |
Specification Test Results
Appearance | Green-gray Powder or pearls |
Titration Complexometric | ≥96.0 % |
Sulfate (SO4) | ≤500 ppm |
Iron (Fe) | ≤400 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...