Mercury(II) bromide 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercuric bromide, Mercury dibromide

AC19049-1000 AC19049-5000
TS19049-1000EA 75 USD
TS19049-1000 TS19049-5000
Mercury(II) bromide 99+%
Mercury(II) bromide
Formula: Br₂Hg
MW: 360.4 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 236 °C
Storage Temperature: Ambient
MDL Number: MFCD00011040
CAS Number: 7789-47-1
EINECS: 232-169-3
UN: 1634
ADR: 6.1,II
Merck Index: 13,05900

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay ≥99.0 %
Chloride (Cl) ≤2000 ppm
Iron (Fe) ≤50 ppm
Insoluble matter ≤0.05 % (in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR