Copper(I) bromide 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

AC19958-0010 AC19958-2500
TS19958-0010EA 323 USD
TS19958-0010 TS19958-2500
Copper(I) bromide 98%, Extra Pure
Copper(I) bromide
Formula: BrCu
Boiling Pt: 1345 °C (1013 hPa)
Melting Pt: 504 °C
Density: 4.71 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010969
CAS Number: 7787-70-4
EINECS: 232-131-6
Merck Index: 13,02689

Order Now

Specification Test Results

Appearance Green to gray-green Powder (May discolor to gray on storage)
Titration Complexometric ≥97.5 %
Sulfate (SO4) ≤5000 ppm
Arsenic (As) ≤50 ppm
Iron (Fe) ≤500 ppm
Insoluble matter ≤0.05 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR