Boron tribromide 99.9%

Supplier: Thermo Scientific Chemicals
Danger

AC29520-0250 AC29520-1000 AC29520-5000
TS29520-0250EA 71 USD
TS29520-0250 TS29520-1000 TS29520-5000
Boron tribromide 99.9%
Boron tribromide
Formula: BBr₃
MW: 250.52 g/mol
Boiling Pt: 91.2
Melting Pt: -46
Density: 2.65 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8 I
Merck Index: 13,01337

Order Now

Specification Test Results

Appearance Clear amber Liquid
Assay ≥99.85 % (By difference)
Titration ≥99.2 %
Sulfur (S) ≤100 ppm
Heavy metals (as Pb) ≤100 ppm
Impurity ≤0.13 % Free bromine (Br2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR