Diphenylsilane 97%

Supplier: Thermo Scientific Chemicals
Warning

AC32977-0050 AC32977-0250
TS32977-0050EA 39 USD
TS32977-0050 TS32977-0250
Diphenylsilane 97%
Diphenylsilane
Formula: C₁₂H₁₂Si
MW: 184.31 g/mol
Boiling Pt: 95
Density: 0.99 g/cm³
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00003002
CAS Number: 775-12-2
EINECS: 212-271-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5780 to 1.5800 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR