Manganese(II) sulfate monohydrate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate

AC20590-0010 AC20590-0050 AC20590-5000
TS20590-0010EA 90 USD
TS20590-0010 TS20590-0050 TS20590-5000
Manganese(II) sulfate monohydrate 99+%, Extra Pure
Manganese(II) sulphate monohydrate
Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700 °C
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 13,05763

Order Now

Specification Test Results

Appearance White to light pink Powder
Titration Complexometric 99.0 to 101.0 %
Loss on ignition 10.0 % to 12.0 % (500°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR