Copper(II) sulfate pentahydrate 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

AC19773-0010 AC19773-0050 AC19773-0250
TS19773-0010EA 86 USD
TS19773-0010 TS19773-0050 TS19773-0250
Copper(II) sulfate pentahydrate 98%, Extra Pure
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Melting Pt: 110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Appearance Blue Crystalline powder or crystals
Titration Iodometric ≥97.5 %
Arsenic (As) ≤10 ppm
Iron (Fe) ≤200 ppm
Lead (Pb) ≤125 ppm
Nickel (Ni) ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR