Ammonium iron(II) sulphate hexahydrate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate

AC20137-0010 AC20137-0050 AC20137-0250 AC20137-0251 AC20137-5000
TS20137-0010EA 149 USD
TS20137-0010 TS20137-0050 TS20137-0250 TS20137-0251 TS20137-5000
Ammonium iron(II) sulphate hexahydrate 99+% for analysis
Ammonium iron(II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Melting Pt: 100 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8

Order Now

Specification Test Results

Appearance Light green Crystalline powder or crystals
Titration with KMnO4 ≥99.0 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤30 ppm
Copper (Cu) ≤20 ppm
Manganese (Mn) ≤500 ppm
Zinc (Zn) ≤30 ppm
Ferric iron (Fe 3+) ≤200 ppm (May increase due to inherent oxidation on storage)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR