Aluminium potassium sulfate, anhydrous, pure

Supplier: Thermo Scientific Chemicals

AC45899-0010 AC45899-0025
TS45899-0010EA 66 USD
TS45899-0010 TS45899-0025
Aluminium potassium sulfate, anhydrous, pure
Aluminium potassium sulphate
Formula: AlKO₈S₂
Boiling Pt: 200 °C (760 mmHg)
Melting Pt: 92 °C
Storage Temperature: Ambient
CAS Number: 10043-67-1

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥90 %
Loss on drying ≤1 %
Ammonium (NH4) ≤0.3 %
Iron (Fe) ≤0.02 %
Heavy metals (as Pb) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR