Ammonium sulphate 99%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

AC43438-0010 AC43438-0050
TS43438-0010EA 56 USD
TS43438-0010 TS43438-0050
Ammonium sulphate 99%, extra pure
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Merck Index: 13,00559

Order Now

Specification Test Results

Appearance White Crystals
Sulfated ash ≤0.05 %
pH 5.0 to 6.0
Titration excess NaOH ≥98.5 %
Chloride (Cl) ≤0.003 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR