Cadmium (II) sulfate 8/3-hydrate, crystals ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cadmium sulphate, 8/3-hydrate, Cadmium sulphate octahydrate
AC40376-0025
AC40376-1000
AC40376-5000
TS40376-0025EA
746
USD
TS40376-0025
TS40376-1000
TS40376-5000
Cadmium (II) sulfate 8/3-hydrate, crystals ACS
Cadmium(II) sulphate 8/3-hydrate
Formula:
3CdSO₄·8H₂O MW: 256.51 g/mol (anhy) Melting Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD02253000 CAS Number: 7790-84-3 EINECS: 233-331-6 UN: 2570 ADR: 6.1,III Merck Index: 13,01627 |
Specification Test Results
Appearance | White Crystals |
Titration Complexometric | 98.0 to 102.0 % |
Chloride (Cl) | ≤0.001 % |
Nitrate and Nitrite (as NO3) | ≤0.003 % |
Calcium (Ca) | ≤0.005 % |
Copper (Cu) | ≤0.002 % |
Iron (Fe) | ≤0.001 % |
Lead (Pb) | ≤0.003 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.02 % |
Zinc (Zn) | ≤0.05 % |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...