Cerium(IV) sulfate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC40434-0250
AC40434-1000
TS40434-0250EA
51
USD
TS40434-0250
TS40434-1000
Cerium(IV) sulfate 99%
Cerium (IV) sulphate
Formula:
Ce(SO₄)₂ MW: 332.24 g/mol Melting Pt: 195 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00148852 CAS Number: 13590-82-4 UN: 3260 ADR: 8,II |
Specification Test Results
Appearance | Yellow Powder and/or chunks |
Titration Iodometric | 39.6 to 44.7 % (Ce) |
Refractive index | 1.5500 to 1.5540 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...