Ammonium sulfamate ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium sulphamidate, Ammonium amidosulphonate

AC42339-0025 AC42339-0050 AC42339-1000 AC42339-5000
TS42339-0025EA 363 USD
TS42339-0025 TS42339-0050 TS42339-1000 TS42339-5000
Ammonium sulfamate ACS
Ammonium sulphamate
Formula: H₆N₂O₃S
MW: 114.13 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00011429
CAS Number: 7773-06-0
EINECS: 231-871-7
Merck Index: 12,00589

Order Now

Specification Test Results

Appearance White Crystalline powder
Melting point 132.0 to 134.0°C
Residue after ignition ≤0.10 %
Titration ≥98.0 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR