Ammonium hexafluorosilicate 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate

AC19400-0100 AC19400-0500 AC19400-2500
TS19400-0100EA 55 USD
TS19400-0100 TS19400-0500 TS19400-2500
Ammonium hexafluorosilicate 99.999% (trace metals basis)
Ammonium hexafluorosilicate
Formula: H₈F₆N₂Si
MW: 178.15
Density: 2.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010887
CAS Number: 16919-19-0
UN: 2854
ADR: 6.1 III
Merck Index: 13,00529

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Total trace metal impurities ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR